Assay Details
Target Gene Details
Entrez Gene ID: | 24000 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 21 |
Gene Aliases: |
PTPD1, PTPRL10 |
Location: |
Chr.12:98676741-98737618 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn21 | NM_001146199.1 | 17 | 3263 | NP_001139671.1 |
| NM_011877.2 | 17 | 3398 | NP_036007.2 | |
| BC055942.1 | 5 | 1787 | AAH55942.1 | |
| BC066856.1 | 15 | 1657 | AAH66856.1 | |
| BC098500.1 | 17 | 3263 | AAH98500.1 | |
| D37801.1 | 17 | 3347 | BAA07053.1 | |
| D83072.1 | 4 | 659 | BAA19740.1 |
More Information
Set Membership: |
|

Genomic Map