Assay Details
Target Gene Details
Entrez Gene ID: | 70948 |
Gene Name: | WD repeat domain 20, retrogene |
Gene Aliases: |
4921538B03Rik, 4930427E19Rik, Wdr20, Wdr20b |
Location: |
Chr.12:65225517-65228454 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr20rt | NM_027614.1 | 2 | 2109 | NP_081890.1 |
| AK015014.1 | 2 | 2109 | BAB29671.1 | |
| BC050778.1 | 2 | 1896 | AAH50778.1 |

Genomic Map