Assay Details
Target Gene Details
Entrez Gene ID: | 68558 |
Gene Name: | ankyrin repeat, family A (RFXANK-like), 2 |
Gene Aliases: |
1110004M18Rik, Ankra |
Location: |
Chr.13:98263076-98274754 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankra2 | NM_001271388.1 | NP_001258317.1 | ||
| NM_001271389.1 | NP_001258318.1 | |||
| NM_001271390.1 | NP_001258319.1 | |||
| NM_001271391.1 | NP_001258320.1 | |||
| NM_001271392.1 | NP_001258321.1 | |||
| NM_023472.2 | NP_075961.1 | |||
| AB221648.1 | ||||
| AF314031.1 | AAK01620.1 | |||
| AK003438.2 | ||||
| AK054037.1 | ||||
| AK081695.1 | BAC38296.1 | |||
| AK139343.1 | BAE23967.1 | |||
| AK161446.1 | ||||
| AK179716.1 | ||||
| BC066113.1 | AAH66113.1 | |||
| BC079615.1 | AAH79615.1 | |||
| BF730624.1 |
More Information
Set Membership: |
|

Genomic Map