Assay Details
Target Gene Details
Entrez Gene ID: | 71853 |
Gene Name: | protein disulfide isomerase associated 6 |
Gene Aliases: |
1700015E05Rik, AL023058, C77895, CaBP5, P5, Txndc7 |
Location: |
Chr.12:17266595-17284770 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdia6 | NM_027959.3 | NP_082235.1 | ||
| AK005989.1 | BAB24354.1 | |||
| AK076558.1 | BAC36392.1 | |||
| AK133792.1 | ||||
| AK151008.1 | ||||
| AK152960.1 | BAE31623.1 | |||
| AK165877.1 | BAE38431.1 | |||
| AK166111.1 | BAE38578.1 | |||
| AK167386.1 | BAE39477.1 | |||
| AK168136.1 | BAE40104.1 | |||
| AK168279.1 | BAE40225.1 | |||
| AK179393.1 | ||||
| BC006865.1 | AAH06865.2 |
More Information
Set Membership: |
|

Genomic Map