Assay Details
Target Gene Details
Entrez Gene ID: | 76260 |
Gene Name: | tetratricopeptide repeat domain 8 |
Gene Aliases: |
0610012F22Rik, AV001447, BBS8 |
Location: |
Chr.12:98920574-98983250 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc8 | NM_029553.3 | 1 | 141 | NP_083829.1 |
| NM_198311.1 | 1 | 103 | NP_938053.1 | |
| AK002597.1 | 1 | 127 | BAB22218.1 | |
| AK032748.1 | 1 | 110 | ||
| AK081697.1 | 1 | 108 | BAC38298.1 | |
| AK144409.1 | 1 | 306 | ||
| BC017523.1 | 1 | 103 | AAH17523.1 | |
| BY144447.1 | 1 | 142 |

Genomic Map