Assay Details
Target Gene Details
Entrez Gene ID: | 382562 |
Gene Name: | profilin family, member 4 |
Gene Aliases: |
2900024P18Rik |
Location: |
Chr.12:4768913-4778808 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pfn4 | NM_028376.3 | 2 | 1039 | NP_082652.1 |
| AK013595.1 | 2 | 1041 | BAB28919.1 | |
| AY495949.1 | 3 | 582 | AAS75143.1 | |
| BC116888.1 | 2 | 883 | ||
| BC116890.1 | 3 | 470 |
More Information
Set Membership: |
|

Genomic Map