Assay Details
Target Gene Details
Entrez Gene ID: | 76260 |
Gene Name: | tetratricopeptide repeat domain 8 |
Gene Aliases: |
0610012F22Rik, AV001447, BBS8 |
Location: |
Chr.12:98920574-98983250 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc8 | NM_029553.3 | NP_083829.1 | ||
| NM_198311.1 | NP_938053.1 | |||
| AK002597.1 | BAB22218.1 | |||
| AK032748.1 | ||||
| AK081697.1 | BAC38298.1 | |||
| BC017523.1 | AAH17523.1 |

Genomic Map