Assay Details
Target Gene Details
Entrez Gene ID: | 69480 |
Gene Name: | tetratricopeptide repeat domain 9 |
Gene Aliases: |
1700029M07Rik, AI429215, AI853559 |
Location: |
Chr.12:81629771-81664941 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc9 | NM_001033149.2 | NP_001028321.1 | ||
| AK133456.1 | BAE21667.1 | |||
| AK161499.1 | BAE36425.1 | |||
| BC028891.1 | AAH28891.1 | |||
| BC139242.1 | ||||
| BC139244.1 |
More Information
Set Membership: |
|

Genomic Map