Assay Details
Target Gene Details
Entrez Gene ID: | 210044 |
Gene Name: | adenylate cyclase 2 |
Gene Aliases: |
mKIAA1060 |
Location: |
Chr.13:68620043-68999541 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcy2 | NM_153534.2 | 5 | 1054 | NP_705762.2 |
| AK122432.1 | 5 | 869 | BAC65714.1 | |
| AK132312.1 | 5 | 1055 | BAE21097.1 | |
| BC037107.1 | 5 | 811 | AAH37107.1 |

Genomic Map