Assay Details
Target Gene Details
Entrez Gene ID: | 30955 |
Gene Name: | phosphoinositide-3-kinase, catalytic, gamma polypeptide |
Gene Aliases: |
5830428L06Rik, PI3Kgamma, p110gamma |
Location: |
Chr.12:32173129-32208677 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3cg | NM_001146200.1 | 10 | 3094 | NP_001139672.1 |
| NM_001146201.1 | 10 | 3150 | NP_001139673.1 | |
| NM_020272.2 | 10 | 3271 | NP_064668.2 | |
| AF208345.1 | 9 | 2950 | AAG41122.1 | |
| AJ249280.1 | 10 | 3271 | CAB89686.1 | |
| AK040543.1 | 10 | 3056 | ||
| AK077772.1 | 4 | 426 | BAC37000.2 | |
| AK132553.1 | 10 | 3096 | BAE21231.1 | |
| AK137470.1 | 10 | 3003 | BAE23366.1 | |
| AK137620.1 | 10 | 3152 | ||
| AK143238.1 | 7 | 775 | BAE25321.1 | |
| BC051246.1 | 10 | 3011 | AAH51246.1 |
More Information
Set Membership: |
|

Genomic Map