Assay Details
Target Gene Details
Entrez Gene ID: | 30955 |
Gene Name: | phosphoinositide-3-kinase, catalytic, gamma polypeptide |
Gene Aliases: |
5830428L06Rik, PI3Kgamma, p110gamma |
Location: |
Chr.12:32173129-32208677 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3cg | NM_001146200.1 | 6 | 2615 | NP_001139672.1 |
| NM_001146201.1 | 6 | 2671 | NP_001139673.1 | |
| NM_020272.2 | 6 | 2792 | NP_064668.2 | |
| AF208345.1 | 5 | 2471 | AAG41122.1 | |
| AJ249280.1 | 6 | 2792 | CAB89686.1 | |
| AK040543.1 | 6 | 2577 | ||
| AK132553.1 | 6 | 2617 | BAE21231.1 | |
| AK137470.1 | 6 | 2524 | BAE23366.1 | |
| AK137620.1 | 6 | 2673 | ||
| AK143238.1 | 3 | 296 | BAE25321.1 | |
| AY831679.1 | 2 | 156 | AAX76838.1 | |
| BC051246.1 | 6 | 2532 | AAH51246.1 |
More Information
Set Membership: |
|

Genomic Map