Assay Details
Target Gene Details
Entrez Gene ID: | 27052 |
Gene Name: | acyloxyacyl hydrolase |
Gene Aliases: |
4930433E13Rik |
Location: |
Chr.13:20794103-21036617 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aoah | NM_001281854.1 | 4 | 608 | NP_001268783.1 |
| NM_012054.4 | 4 | 608 | NP_036184.1 | |
| AK084452.1 | 4 | 603 | BAC39187.1 | |
| AK085343.1 | 4 | 578 | ||
| AK133301.1 | 4 | 549 | BAE21600.1 | |
| AK150886.1 | 4 | 584 | BAE29933.1 | |
| AK152346.1 | 4 | 584 | BAE31141.1 | |
| AK170293.1 | 3 | 144 | BAE41693.1 | |
| AK171572.1 | 4 | 427 | BAE42533.1 | |
| BC089580.1 | 4 | 588 | AAH89580.1 | |
| BC150767.1 | 4 | 402 | ||
| CT010242.1 | 4 | 327 | CAJ18450.1 |
More Information
Set Membership: |
|

Genomic Map