Assay Details
Target Gene Details
Entrez Gene ID: | 104759 |
Gene Name: | phospholipase D family, member 4 |
Gene Aliases: |
AI132321, thss |
Location: |
Chr.12:112760655-112768988 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pld4 | NM_178911.4 | 8 | 1088 | NP_849242.1 |
| AK045459.1 | 8 | 1047 | BAC32379.1 | |
| AK079549.1 | 8 | 1047 | BAC37681.1 | |
| AK079878.1 | 8 | 1090 | BAC37771.1 | |
| AK151551.1 | 8 | 1322 | BAE30496.1 | |
| AK152969.1 | 7 | 936 | ||
| AK170362.1 | 8 | 1045 | BAE41745.1 | |
| BC058565.1 | 8 | 1038 | AAH58565.1 |
More Information
Set Membership: |
|

Genomic Map