Assay Details
Target Gene Details
Entrez Gene ID: | 22018 |
Gene Name: | thyroid peroxidase |
Gene Aliases: |
- |
Location: |
Chr.12:30054661-30132984 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpo | NM_009417.2 | 12 | 2190 | NP_033443.1 |
| AK047843.1 | 12 | 2169 | BAC33171.1 | |
| X60703.1 | 12 | 2190 | CAA43114.1 |
More Information
Set Membership: |
|

Genomic Map