Assay Details
Target Gene Details
Entrez Gene ID: | 217847 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10 |
Gene Aliases: |
PZI, ZPI |
Location: |
Chr.12:103616675-103631490 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina10 | NM_001301404.1 | NP_001288333.1 | ||
| NM_144834.4 | NP_659083.2 | |||
| AY324633.1 | AAP87100.1 | |||
| BC018416.1 | AAH18416.1 |
More Information
Set Membership: |
|

Genomic Map