Assay Details
Target Gene Details
Entrez Gene ID: | 74413 |
Gene Name: | tandem C2 domains, nuclear |
Gene Aliases: |
4933406D09Rik, Mtac2d1, Tac2-N |
Location: |
Chr.12:101645446-101719000 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tc2n | NM_001082976.2 | 11 | 1380 | NP_001076445.1 |
| NM_001286364.1 | 11 | 2043 | NP_001273293.1 | |
| NM_028924.3 | 11 | 1732 | NP_083200.1 | |
| AB062282.1 | 10 | 1212 | BAB62893.1 | |
| AK016683.1 | 11 | 2064 | BAB30378.1 | |
| AK136558.1 | 11 | 1381 | BAE23043.1 | |
| BC120793.1 | 11 | 1732 |

Genomic Map