Assay Details
Target Gene Details
Entrez Gene ID: | 74413 |
Gene Name: | tandem C2 domains, nuclear |
Gene Aliases: |
4933406D09Rik, Mtac2d1, Tac2-N |
Location: |
Chr.12:101645446-101719000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tc2n | NM_001082976.2 | NP_001076445.1 | ||
| NM_001286364.1 | NP_001273293.1 | |||
| NM_028924.3 | NP_083200.1 | |||
| AB062282.1 | BAB62893.1 | |||
| AK016683.1 | BAB30378.1 | |||
| AK136558.1 | BAE23043.1 | |||
| BC120793.1 |

Genomic Map