Assay Details
Target Gene Details
Entrez Gene ID: | 11624 |
Gene Name: | aryl-hydrocarbon receptor repressor |
Gene Aliases: |
mKIAA1234 |
Location: |
Chr.13:74211117-74292330 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahrr | NM_009644.2 | NP_033774.1 | ||
| AB015140.1 | BAA37132.1 | |||
| AK154152.1 | BAE32410.1 | |||
| AK154761.1 | BAE32810.1 | |||
| AK155711.1 | BAE33396.1 | |||
| AK173134.1 | BAD32412.1 | |||
| BC118534.1 |
More Information
Set Membership: |
|

Genomic Map