Assay Details
Target Gene Details
Entrez Gene ID: | 20773 |
Gene Name: | serine palmitoyltransferase, long chain base subunit 2 |
Gene Aliases: |
AI173915, LCB2, mKIAA0526 |
Location: |
Chr.12:87307889-87388230 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptlc2 | NM_011479.3 | 1 | 92 | NP_035609.1 |
| AK008871.1 | 1 | 91 | ||
| AK044917.1 | 1 | 93 | BAC32140.1 | |
| AK057644.1 | 1 | 128 | ||
| AK089019.1 | 1 | 86 | BAC40704.1 | |
| AK159346.1 | 1 | 93 | BAE35008.1 | |
| BC003227.1 | 1 | 125 | AAH03227.1 | |
| U27455.1 | 1 | 60 | AAC53310.1 | |
| X95642.1 | 1 | 63 | CAA64898.1 |
More Information
Set Membership: |
|

Genomic Map