Assay Details
Target Gene Details
Entrez Gene ID: | 20773 |
Gene Name: | serine palmitoyltransferase, long chain base subunit 2 |
Gene Aliases: |
AI173915, LCB2, mKIAA0526 |
Location: |
Chr.12:87307889-87388230 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sptlc2 | NM_011479.3 | 11 | 1558 | NP_035609.1 |
| AK042149.1 | 1 | 72 | BAC31185.1 | |
| AK044917.1 | 11 | 1559 | BAC32140.1 | |
| AK057644.1 | 11 | 1594 | ||
| AK080181.1 | 5 | 1756 | BAC37842.1 | |
| AK089019.1 | 11 | 1552 | BAC40704.1 | |
| AK133728.1 | 5 | 619 | BAE21805.1 | |
| AK139681.1 | 3 | 261 | BAE24102.1 | |
| AK159346.1 | 11 | 1559 | BAE35008.1 | |
| AK172965.1 | 4 | 354 | BAD32243.1 | |
| BC003227.1 | 11 | 1591 | AAH03227.1 | |
| U27455.1 | 11 | 1526 | AAC53310.1 | |
| X95642.1 | 11 | 1529 | CAA64898.1 |
More Information
Set Membership: |
|

Genomic Map