Assay Details
Target Gene Details
Entrez Gene ID: | 52690 |
Gene Name: | SET domain containing 3 |
Gene Aliases: |
2610102I01Rik, 2610305M23Rik, 6030490B17Rik, D12Ertd771e |
Location: |
Chr.12:108106431-108179381 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setd3 | NM_028262.3 | NP_082538.2 | ||
| AK011993.1 | BAB27964.1 | |||
| AK029403.1 | BAC26435.1 | |||
| AK031371.1 | BAC27371.1 | |||
| AK146777.1 | BAE27425.1 | |||
| AK166570.1 | BAE38861.1 | |||
| AK170917.1 | 1 | 111 | ||
| AY513271.1 | AAS82953.1 | |||
| BC016123.1 | AAH16123.1 | |||
| BC019973.1 | AAH19973.1 | |||
| BC057968.1 | AAH57968.1 | |||
| BY087049.1 |
More Information
Set Membership: |
|

Genomic Map