Assay Details
Target Gene Details
Entrez Gene ID: | 109929 |
Gene Name: | zinc finger and BTB domain containing 25 |
Gene Aliases: |
2810462M08Rik, 2900064P18Rik, AI842128, Kup, Zfp-50, Zfp50 |
Location: |
Chr.12:76348900-76370218 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb25 | NM_001172104.1 | NP_001165575.1 | ||
| NM_028356.2 | NP_082632.2 | |||
| AK137486.1 | ||||
| BC068249.1 | AAH68249.1 |
More Information
Set Membership: |
|

Genomic Map