Assay Details
Target Gene Details
Entrez Gene ID: | 59050 |
Gene Name: | NSA2 ribosome biogenesis homolog (S. cerevisiae) |
Gene Aliases: |
5730427N09Rik, LNR42, Nsa2p, Tinp1, Yr29 |
Location: |
Chr.13:97129427-97137926 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsa2 | NM_021552.5 | NP_067527.3 | ||
| AF238866.1 | AAF63492.1 | |||
| AK002474.1 | BAB22128.2 | |||
| AK009136.1 | ||||
| AK012832.1 | BAB28500.1 | |||
| AK014275.1 | BAB29237.1 | |||
| AK017605.1 | BAB30834.1 | |||
| AK078457.1 | BAC37283.1 | |||
| BC086776.1 | AAH86776.1 | |||
| BC108365.1 | AAI08366.1 | |||
| BC126976.1 |

Genomic Map