Assay Details
Target Gene Details
Entrez Gene ID: | 67452 |
Gene Name: | patatin-like phospholipase domain containing 8 |
Gene Aliases: |
1200006O19Rik, AI467579, Ipla2(gamma) |
Location: |
Chr.12:44269128-44315935 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnpla8 | NM_026164.2 | NP_080440.2 | ||
| AB044139.1 | BAB97200.1 | |||
| AK004621.1 | BAB23417.1 | |||
| AK017838.1 | ||||
| AK145776.1 | BAE26645.1 | |||
| AK163211.1 | BAE37236.1 | |||
| AK168475.1 | BAE40365.1 | |||
| BC019364.1 | AAH19364.1 | |||
| BC127056.1 |
More Information
Set Membership: |
|

Genomic Map