Assay Details
Target Gene Details
Entrez Gene ID: | 19363 |
Gene Name: | RAD51 homolog B |
Gene Aliases: |
AI553500, R51H2, Rad51l1, mREC2 |
Location: |
Chr.12:79297282-79852895 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rad51b | NM_001252562.1 | NP_001239491.1 | ||
| NM_009014.3 | NP_033040.2 | |||
| AK015771.1 | BAB29970.1 | |||
| AK032913.1 | BAC28084.1 | |||
| U92068.1 | AAB63359.1 |
More Information
Set Membership: |
|

Genomic Map