Assay Details
Target Gene Details
Entrez Gene ID: | 94186 |
Gene Name: | striatin, calmodulin binding protein 3 |
Gene Aliases: |
Gs2na, SG2NA |
Location: |
Chr.12:51608541-51692565 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Strn3 | NM_001172098.1 | NP_001165569.1 | ||
| NM_052973.2 | NP_443205.1 | |||
| AF307777.1 | AAG24454.1 | |||
| AK139183.1 | BAE23914.1 | |||
| AK140447.1 | ||||
| AK165908.1 | ||||
| BC138055.1 | ||||
| BC145127.1 | ||||
| EF685152.1 | ||||
| EF685153.1 | ||||
| EF685154.1 | ||||
| EF685155.1 |

Genomic Map