Assay Details
Target Gene Details
Entrez Gene ID: | 74691 |
Gene Name: | tudor domain containing 9 |
Gene Aliases: |
4930441E05Rik |
Location: |
Chr.12:111971474-112068854 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tdrd9 | NM_029056.1 | 4 | 611 | NP_083332.1 |
| AB362563.1 | 4 | 611 | ||
| BC115831.1 | 1 | 59 | ||
| BC116656.1 | 1 | 59 |
More Information
Set Membership: |
|

Genomic Map