Assay Details
Target Gene Details
Entrez Gene ID: | 14186 |
Gene Name: | fibroblast growth factor receptor 4 |
Gene Aliases: |
Fgfr-4 |
Location: |
Chr.13:55152818-55168759 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgfr4 | NM_008011.2 | NP_032037.2 | ||
| AK084850.1 | BAC39292.1 | |||
| AK085723.1 | ||||
| AY493377.2 | AAS72387.2 | |||
| BC033313.1 | AAH33313.1 | |||
| DQ388428.1 | ABD43187.1 | |||
| X59927.1 | CAA42551.1 |
More Information
Set Membership: |
|

Genomic Map