Assay Details
Target Gene Details
Entrez Gene ID: | 14186 |
Gene Name: | fibroblast growth factor receptor 4 |
Gene Aliases: |
Fgfr-4 |
Location: |
Chr.13:55152818-55168759 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgfr4 | NM_008011.2 | 6 | 815 | NP_032037.2 |
| AK084850.1 | 6 | 815 | BAC39292.1 | |
| AK085723.1 | 6 | 814 | ||
| AY493377.2 | 5 | 701 | AAS72387.2 | |
| BC033313.1 | 6 | 1002 | AAH33313.1 | |
| DQ388428.1 | 5 | 701 | ABD43187.1 | |
| X59927.1 | 6 | 757 | CAA42551.1 |
More Information
Set Membership: |
|

Genomic Map