Assay Details
Target Gene Details
Entrez Gene ID: | 16198 |
Gene Name: | interleukin 9 |
Gene Aliases: |
Il-9, P40 |
Location: |
Chr.13:56479277-56482246 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il9 | NM_008373.1 | NP_032399.1 | ||
| X14045.1 | CAA32203.1 |
More Information
Set Membership: |
|

Genomic Map