Assay Details
Target Gene Details
Entrez Gene ID: | 13361 |
Gene Name: | dihydrofolate reductase |
Gene Aliases: |
8430436I03Rik, AA607882, AI662710, AW555094 |
Location: |
Chr.13:92354783-92389053 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhfr | NM_010049.3 | NP_034179.1 | ||
| AK031241.1 | BAC27315.1 | |||
| AK136212.1 | BAE22879.1 | |||
| BC005796.1 | AAH05796.1 | |||
| L26316.1 | AAA37523.1 | |||
| M10811.1 | AAA37525.1 | |||
| V00734.1 | CAA24112.1 | |||
| X56066.1 | CAA39544.1 |
More Information
Set Membership: |
|

Genomic Map