Assay Details
Target Gene Details
Entrez Gene ID: | 14062 |
Gene Name: | coagulation factor II (thrombin) receptor |
Gene Aliases: |
AI482343, Cf2r, Par1, ThrR |
Location: |
Chr.13:95601789-95618433 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2r | NM_010169.3 | NP_034299.2 | ||
| AK085990.1 | 1 | 36 | BAC39587.1 | |
| AK146194.1 | BAE26968.1 | |||
| AK159282.1 | 1 | 50 | BAE34960.1 | |
| AK160032.1 | 1 | 50 | BAE35575.1 | |
| BC031516.1 | AAH31516.1 | |||
| L03529.1 | AAA40438.1 |
More Information
Set Membership: |
|

Genomic Map