Assay Details
Target Gene Details
Entrez Gene ID: | 14063 |
Gene Name: | coagulation factor II (thrombin) receptor-like 1 |
Gene Aliases: |
Gpcr11, PAR-2, Par2 |
Location: |
Chr.13:95511718-95525240 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F2rl1 | NM_007974.4 | NP_032000.3 | ||
| AK144986.1 | BAE26169.1 | |||
| AK158462.1 | BAE34521.1 | |||
| AK160916.1 | BAE36090.1 | |||
| AX785009.1 | CAE11955.1 | |||
| BC025432.1 | AAH25432.1 | |||
| BY029876.1 | ||||
| Z48043.1 | CAA88097.1 |
More Information
Set Membership: |
|

Genomic Map