Assay Details
Target Gene Details
Entrez Gene ID: | 75805 |
Gene Name: | neurolysin (metallopeptidase M3 family) |
Gene Aliases: |
4930472G13Rik, C79345 |
Location: |
Chr.13:104023050-104109660 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nln | NM_029447.2 | 12 | 2029 | NP_083723.1 |
| AK015561.1 | 1 | 38 | ||
| AK050120.1 | 2 | 143 | BAC34076.1 | |
| AK079090.1 | 12 | 2030 | BAC37533.1 | |
| AK138399.1 | 12 | 2002 | BAE23644.1 | |
| AK146364.1 | 12 | 1978 | BAE27114.1 | |
| AK170508.1 | 12 | 2030 | ||
| AK171432.1 | 12 | 2030 | BAE42449.1 | |
| BC016224.1 | 12 | 2002 | AAH16224.1 | |
| BC025520.1 | 5 | 888 | AAH25520.1 |
More Information
Set Membership: |
|

Genomic Map