Assay Details
Target Gene Details
Entrez Gene ID: | 218236 |
Gene Name: | family with sequence similarity 120, member A |
Gene Aliases: |
C9orf10, Ossa |
Location: |
Chr.13:48879216-48967877 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam120a | NM_001033268.2 | 11 | 1988 | NP_001028440.2 |
| AK172909.1 | 11 | 2177 | BAD32187.1 | |
| BC010304.1 | 7 | 1056 | AAH10304.1 | |
| BC042582.1 | 10 | 1504 | ||
| BC158134.1 | 11 | 1980 |

Genomic Map