Assay Details
Target Gene Details
Entrez Gene ID: | 380836 |
Gene Name: | MRS2 magnesium homeostasis factor homolog (S. cerevisiae) |
Gene Aliases: |
Gm902, HPT, Mrs2l, RPT |
Location: |
Chr.13:24987483-25020358 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrs2 | NM_001013389.2 | 10 | 1209 | NP_001013407.2 |
| AK136100.1 | 5 | 500 | ||
| BB015377.2 | 3 | 310 |
More Information
Set Membership: |
|

Genomic Map