Assay Details
Target Gene Details
Entrez Gene ID: | 17686 |
Gene Name: | mutS homolog 3 (E. coli) |
Gene Aliases: |
D13Em1, Rep-3, Rep3 |
Location: |
Chr.13:92211879-92355003 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msh3 | NM_010829.2 | NP_034959.2 | ||
| AK083388.1 | ||||
| AK133907.1 | ||||
| AK161089.1 | ||||
| BC150759.1 | ||||
| BQ571929.1 | ||||
| M80360.1 | AAA40052.1 |
More Information
Set Membership: |
|

Genomic Map