Assay Details
Target Gene Details
Entrez Gene ID: | 72562 |
Gene Name: | pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 2 |
Gene Aliases: |
2700061N24Rik, Dcoh2, Dcohm |
Location: |
Chr.13:55727124-55776830 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcbd2 | NM_028281.1 | NP_082557.1 | ||
| AA013784.1 | ||||
| AK012465.1 | BAB28260.1 | |||
| BC028642.1 | AAH28642.1 | |||
| BY157602.1 |
More Information
Set Membership: |
|

Genomic Map