Assay Details
Target Gene Details
Entrez Gene ID: | 67252 |
Gene Name: | CAP, adenylate cyclase-associated protein, 2 (yeast) |
Gene Aliases: |
2810452G09Rik, AV045235 |
Location: |
Chr.13:46501903-46650281 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cap2 | NM_026056.4 | 11 | 1472 | NP_080332.1 |
| AK010235.1 | 11 | 1219 | BAB26786.1 | |
| AK013331.1 | 11 | 1229 | BAB28795.1 | |
| AK014979.1 | 7 | 867 | ||
| AK033071.1 | 11 | 1228 | BAC28143.1 | |
| AK035556.1 | 11 | 1473 | BAC29104.1 | |
| AK047701.1 | 9 | 1039 | BAC33131.1 | |
| AK049813.1 | 8 | 915 | ||
| AK076737.1 | 7 | 892 | ||
| BC050752.1 | 11 | 1233 | AAH50752.2 | |
| BC057937.1 | 11 | 1287 | AAH57937.1 |
More Information
Set Membership: |
|

Genomic Map