Assay Details
Target Gene Details
Entrez Gene ID: | 75678 |
Gene Name: | inositol 1,3,4,5,6-pentakisphosphate 2-kinase |
Gene Aliases: |
1810043M15Rik, InsP6 |
Location: |
Chr.13:49421223-49463060 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ippk | NM_001276399.1 | NP_001263328.1 | ||
| NM_199056.3 | NP_951011.1 | |||
| NR_075085.1 | ||||
| AK029085.1 | BAE20447.1 | |||
| AK031987.1 | ||||
| AK160092.1 | BAE35623.1 | |||
| AK167467.1 | BAE39551.1 | |||
| BB619387.1 | ||||
| BC024630.1 | ||||
| BC062167.1 | AAH62167.1 | |||
| BC065157.1 | AAH65157.1 |
More Information
Set Membership: |
|

Genomic Map