Assay Details
Target Gene Details
Entrez Gene ID: | 110789 |
Gene Name: | adhesion G protein-coupled receptor V1 |
Gene Aliases: |
Frings, Gpr98, Mass1, Mgr1, VLGR1 |
Location: |
Chr.13:81095068-81633197 on Build GRCm38 |
Assay Gene Location: | Within Exon 68 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgrv1 | NM_054053.4 | 68 | 13916 | NP_473394.3 |
| AF435926.1 | 68 | 13917 | AAL30812.1 | |
| AK129190.2 | 5 | 885 | BAC98000.3 |
More Information
Set Membership: |
|

Genomic Map