Assay Details
Target Gene Details
Entrez Gene ID: | 17686 |
Gene Name: | mutS homolog 3 (E. coli) |
Gene Aliases: |
D13Em1, Rep-3, Rep3 |
Location: |
Chr.13:92211879-92355003 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msh3 | NM_010829.2 | 9 | 1380 | NP_034959.2 |
| AK040616.1 | 7 | 3707 | BAC30646.1 | |
| AK083388.1 | 10 | 1693 | ||
| AK133907.1 | 9 | 1372 | ||
| AK161089.1 | 9 | 1376 | ||
| BC150759.1 | 9 | 1299 | ||
| BQ571929.1 | 3 | 386 | ||
| M80360.1 | 9 | 1377 | AAA40052.1 |
More Information
Set Membership: |
|

Genomic Map