Assay Details
Target Gene Details
Entrez Gene ID: | 17755 |
Gene Name: | microtubule-associated protein 1B |
Gene Aliases: |
A230055D22, AI843217, LC1, MAP5, Mtap-5, Mtap1b, Mtap5 |
Location: |
Chr.13:99421464-99516602 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map1b | NM_008634.2 | 2 | 351 | NP_032660.2 |
| AK158307.1 | 2 | 310 | BAE34454.1 | |
| AK165800.1 | 2 | 268 | ||
| BC019183.1 | 2 | 283 | ||
| BC037105.1 | 2 | 296 | AAH37105.1 | |
| BC138033.1 | 2 | 281 | ||
| BC138034.1 | 2 | 281 | ||
| BY003859.1 | 2 | 351 | ||
| X51396.1 | 2 | 289 | CAA35761.1 |
More Information
Set Membership: |
|

Genomic Map