Assay Details
Target Gene Details
Entrez Gene ID: | 12380 |
Gene Name: | calpastatin |
Gene Aliases: |
- |
Location: |
Chr.13:74692366-74808874 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cast | NM_001301153.1 | NP_001288082.1 | ||
| NM_001301155.1 | NP_001288084.1 | |||
| NM_001301156.1 | NP_001288085.1 | |||
| NM_001301157.1 | NP_001288086.1 | |||
| NM_001301158.1 | NP_001288087.1 | |||
| NM_001301160.1 | NP_001288089.1 | |||
| NM_001301181.1 | NP_001288110.1 | |||
| NM_009817.4 | NP_033947.1 | |||
| AB026997.1 | BAA84768.1 | |||
| AF190151.1 | AAF25193.1 | |||
| AF190152.1 | AAF25194.1 | |||
| AK006253.1 | ||||
| AK028841.1 | ||||
| AK028843.1 | BAC26149.1 | |||
| AK029293.1 | BAC26377.1 | |||
| AK036576.1 | BAC29486.1 | |||
| AK040688.1 | BAC30668.2 | |||
| AK083310.1 | ||||
| AK083395.1 | ||||
| AK089108.1 | BAC40755.1 | |||
| AK161284.1 | BAE36293.1 | |||
| BC010603.1 | AAH10603.1 |
More Information
Set Membership: |
|

Genomic Map