Assay Details
Target Gene Details
Entrez Gene ID: | 14780 |
Gene Name: | glutathione peroxidase 5 |
Gene Aliases: |
AV379049, Arep |
Location: |
Chr.13:21286429-21292729 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpx5 | NM_010343.2 | NP_034473.2 | ||
| AK029731.1 | BAC26586.1 | |||
| BC100749.1 | AAI00750.1 | |||
| BC103588.1 | AAI03589.1 | |||
| BC103589.1 | AAI03590.1 | |||
| BC105648.1 |
More Information
Set Membership: |
|

Genomic Map