Assay Details
Target Gene Details
Entrez Gene ID: | 74129 |
Gene Name: | dimethylglycine dehydrogenase precursor |
Gene Aliases: |
1200014D15Rik, AI787269 |
Location: |
Chr.13:93674436-93752823 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dmgdh | NM_028772.3 | NP_083048.1 | ||
| AK004755.1 | BAB23536.1 | |||
| BC024126.1 | AAH24126.1 | |||
| BC089599.1 | AAH89599.1 |
More Information
Set Membership: |
|

Genomic Map