Assay Details
Target Gene Details
Entrez Gene ID: | 80898 |
Gene Name: | endoplasmic reticulum aminopeptidase 1 |
Gene Aliases: |
Arts1, ERAAP, PILSA, PILSAP |
Location: |
Chr.13:74639624-74691931 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erap1 | NM_030711.4 | NP_109636.1 | ||
| AB047552.1 | BAB11982.1 | |||
| AF227511.1 | AAG44260.1 | |||
| AK029841.1 | ||||
| AK030329.1 | BAC26904.1 | |||
| AK040350.1 | BAC30569.1 | |||
| AK159406.1 | ||||
| BC046610.1 | AAH46610.1 |
More Information
Set Membership: |
|

Genomic Map