Assay Details
Target Gene Details
Entrez Gene ID: | 80898 |
Gene Name: | endoplasmic reticulum aminopeptidase 1 |
Gene Aliases: |
Arts1, ERAAP, PILSA, PILSAP |
Location: |
Chr.13:74639624-74691931 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erap1 | NM_030711.4 | 3 | 633 | NP_109636.1 |
| AB047552.1 | 2 | 621 | BAB11982.1 | |
| AF227511.1 | 2 | 609 | AAG44260.1 | |
| AK029841.1 | 3 | 632 | ||
| AK030329.1 | 3 | 634 | BAC26904.1 | |
| AK081181.1 | 3 | 660 | BAC38159.1 | |
| AK159406.1 | 3 | 636 | ||
| AK163954.1 | 3 | 653 | ||
| BC046610.1 | 3 | 639 | AAH46610.1 |
More Information
Set Membership: |
|

Genomic Map