Assay Details
Target Gene Details
Entrez Gene ID: | 80898 |
Gene Name: | endoplasmic reticulum aminopeptidase 1 |
Gene Aliases: |
Arts1, ERAAP, PILSA, PILSAP |
Location: |
Chr.13:74639624-74691931 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erap1 | NM_030711.4 | 6 | 1071 | NP_109636.1 |
| AB047552.1 | 5 | 1059 | BAB11982.1 | |
| AF227511.1 | 5 | 1047 | AAG44260.1 | |
| AK029841.1 | 6 | 1070 | ||
| AK030329.1 | 6 | 1072 | BAC26904.1 | |
| AK040350.1 | 3 | 289 | BAC30569.1 | |
| AK081181.1 | 6 | 1098 | BAC38159.1 | |
| AK159406.1 | 6 | 1072 | ||
| AK163954.1 | 6 | 1091 | ||
| BC046610.1 | 6 | 1077 | AAH46610.1 |
More Information
Set Membership: |
|

Genomic Map