Assay Details
Target Gene Details
Entrez Gene ID: | 18570 |
Gene Name: | programmed cell death 6 |
Gene Aliases: |
AV299538, Alg2, MA-3, PS2, alg-2 |
Location: |
Chr.13:74303121-74317575 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdcd6 | NM_011051.3 | 5 | 547 | NP_035181.1 |
| AK008610.1 | 5 | 471 | BAB25775.1 | |
| AK028223.1 | 5 | 502 | BAC25823.1 | |
| AK078044.1 | 5 | 475 | BAC37107.1 | |
| AK150335.1 | 5 | 465 | BAE29477.1 | |
| BC040079.1 | 5 | 498 | AAH40079.1 | |
| BU604816.1 | 5 | 550 | ||
| U49112.1 | 5 | 484 | AAB38108.1 | |
| X14938.1 | 4 | 314 | CAA33064.1 |
More Information
Set Membership: |
|

Genomic Map